missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) fluoride tetrahydrate, Puratronic™, 99.99% (metals basis)

Product Code. 11350949
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11350949 5 g 5g
11340949 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11350949 Supplier Thermo Scientific Alfa Aesar Supplier No. 010693.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13817-37-3 | CoF2H8O4 | 168.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13817-37-3
Molecular Formula CoF2H8O4
Molecular Weight (g/mol) 168.99
MDL Number MFCD00149656
InChI Key MNHUTFNEBYCZAF-UHFFFAOYSA-L
Synonym cobalt 2+ tetrahydrate difluoride, acmc-1btz4, cobaltous fluoride 4h2o, cobalt ii ; fluoride tetrahydrate, cobalt 2+ fluoride-water 1/2/4, cobalt fluoride cof2 ,tetrahydrate 8ci,9ci
PubChem CID 24884271
SMILES O.O.O.O.[F-].[F-].[Co++]

Specifications

Melting Point 200°C (decomposition)
Color Pink
Physical Form Powder
Quantity 5 g
UN Number UN2923
Merck Index 14,2440
IUPAC Name cobalt(2+);difluoride;tetrahydrate
Formula Weight 168.99 (96.93 Anhydrous)
Percent Purity 99.99%
Grade Puratronic
Odor Odorless
Sensitivity Moisture sensitive
Density 2.19 g/mL
Assay (metals basis)
Chemical Name or Material Cobalt(II) fluoride tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.