missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) fluoride, anhydrous, 98%

Product Code. 11352168
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352168 25 g 25g
11362168 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352168 Supplier Thermo Scientific Alfa Aesar Supplier No. 013074.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10026-17-2 | CoF2 | 96.93 g/mol

Cobalt(II) fluoride can be used as a catalyst to alloy metals. It is also used for optical deposition, of which it tremendously improves optical quality. The compound may be used in dental care.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-17-2
Molecular Formula CoF2
Molecular Weight (g/mol) 96.93
MDL Number MFCD00010941
InChI Key YCYBZKSMUPTWEE-UHFFFAOYSA-L
Synonym cobalt ii fluoride, cobalt fluoride, cobalt difluoride, cobaltous fluoride, hsdb 236, cobalt fluoride pink, cobalt ii fluoride, powder, cobalt ii fluoride, anhydrous, cobalt ii fluoride, ultra dry
PubChem CID 24820
IUPAC Name difluorocobalt
SMILES F[Co]F

Specifications

Boiling Point 1400°C
Melting Point 1200°C
Physical Form Powder
Quantity 25 g
UN Number UN2923
Merck Index 14,2440
Solubility Information Soluble in water and hydrofluoric acid. insoluble in alcohol,ether,benzene.
Formula Weight 96.93
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 4.46 g/mL
Chemical Name or Material Cobalt(II) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.