missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Diacetate, TRC

Product Code. 30356156
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30356156 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356156 Supplier Toronto Research Chemicals Supplier No. TRCC6330951G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cobalt(II) Diacetate
CAS 71-48-7
Molecular Formula C4H6O4 .Co
Quantity 1 g
Synonym Acetic Acid Cobalt Salt, Bis(acetato)cobalt, Cobaltous Acetate, Cobaltous Diacetate, Cobalt Acetate
InChI Formula InChI=1S/2C2H4O2.Co/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2
SMILES [Co+2].CC(=O)[O-].CC(=O)[O-]
IUPAC Name cobalt(2+);diacetate
Molecular Weight (g/mol) 177.02
Formula Weight 176.9598
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.