missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) citrate dihydrate, 98%

Product Code. 11481087
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11481087

Brand: Thermo Scientific Alfa Aesar B24088.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6424-15-3 | C12H20Co3O16 | 597.076 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6424-15-3
Molecular Formula C12H20Co3O16
Molecular Weight (g/mol) 597.076
MDL Number MFCD02684007
InChI Key SKMIIAIULKZNRQ-UHFFFAOYSA-N
Synonym cobalt ii citrate dihydrate
PubChem CID 131882668
IUPAC Name cobalt;2-hydroxypropane-1,2,3-tricarboxylic acid;dihydrate
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O.O.[Co].[Co].[Co]

Specifications

Melting Point ∼150°C (decomposition)
Quantity 25 g
UN Number UN3077
Formula Weight 591.02 (554.99 Anhydrous)
Percent Purity 98%
Chemical Name or Material Cobalt(II) citrate dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.