missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride hexahydrate, 99.9% (metals basis)

Product Code. 11311119
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311119 250 g 250g
11301119 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311119 Supplier Thermo Scientific Alfa Aesar Supplier No. 011344.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sensitivity: Hygroscopic; m.p. 52-56°C -4H2O, 100°C -5H2O, 120-140°C -6H2O | CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol

Cobalt(II) chloride hexahydrate is used as a humidity indicator in weather instruments. It finds use in electroplating of cobalt, in invisible ink, in organic chemistry and is a precursor to cobaltocene(bis (cyclopentadienyl) cobalt(II)).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Melting Point 87°C
Color Purple-Red
Physical Form Crystals
Quantity 250 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information Soluble in water,alcohol,acetone,ether and glycerol.
Formula Weight 237.93 (129.84 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 1.924
Chemical Name or Material Cobalt(II) chloride hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.