Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Color | Lilac |
| Physical Form | Fine Crystalline Powder |
| Quantity | 500 g |
| Assay Percent Range | 98-102% |
| Linear Formula | CoCl2·6H2O |
| Fieser | 01,155; 06,127; 11,479 |
| Merck Index | 15, 2421 |
| Formula Weight | 237.93 |
| Percent Purity | 98.0 to 102.0% |
| Grade | ACS Reagent |
| Show More |
Safety and Handling
- Cobalt(II) chloride hexahydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H341-Suspected of causing genetic defects.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.