missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Chloride Anhydrous, Technical, SLR, Fisher Chemical

Product Code. 10020070 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10020070 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10020070 Supplier Fisher Chemical Supplier No. C/6600/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Specifikationer

Boiling Point 1049°C
Melting Point 735°C
CAS 7646-79-9
Color Blue
pH 4.9
Physical Form Solid
Quantity 100 g
Molecular Formula Cl2Co
MDL Number 10938
InChI Key GVPFVAHMJGGAJG-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Co++]
IUPAC Name λ2-cobalt(2+) dichloride
Molecular Weight (g/mol) 129.83
Formula Weight 129.84g/mol
Packaging Plastic powder jar
Vapor Pressure 40mmHg 770
Visa mer Visa mindre
compliance-icons compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • Cobalt(II) chloride, anhydrous
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Faroangivelser
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H341-Suspected of causing genetic defects.
  • H332-Harmful if inhaled.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.