missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Carbonate Hydrate, TRC

Product Code. 30349264
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349264 5 g 5g
30354401 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30349264 Supplier Toronto Research Chemicals Supplier No. TRCC6330805G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cobalt(II) Carbonate Hydrate
CAS 57454-67-8
Molecular Formula CCoO3 . (H2O)x
Quantity 5 g
Synonym Carbonic Acid Cobalt(2+) Salt (1:1) Hydrate
InChI Formula InChI=1S/CH2O3.2Co.H2O/c2-1(3)4;;;/h(H2,2,3,4);;;1H2/q;;+1;/p-3
SMILES [O-]C([O-])=O.[Co][Co+].[H2O]
IUPAC Name Cobalt(II) Carbonate Hydrate
Molecular Weight (g/mol) 136.96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.