missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) bromide hydrate, Thermo Scientific Chemicals

Catalog Number 11399377
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
85017-77-2
Br2Co
218.74
MFCD00016017
BZRRQSJJPUGBAA-UHFFFAOYSA-L
cobalt ii bromide hydrate, acmc-20alea, br2co.h2o, cobaltous bromide hydrate, dibromocobalt-water 1/1, cobalt ii bromide hexahydrate, cobalt bromide cobr2 ,hydrate 9ci
16212115
[Co++].[Br-].[Br-]
This item is not returnable. View return policy
85017-77-2
Br2Co
218.74
MFCD00016017
BZRRQSJJPUGBAA-UHFFFAOYSA-L
cobalt ii bromide hydrate, acmc-20alea, br2co.h2o, cobaltous bromide hydrate, dibromocobalt-water 1/1, cobalt ii bromide hexahydrate, cobalt bromide cobr2 ,hydrate 9ci
16212115
[Co++].[Br-].[Br-]

For Hygrometers, Catalysts

Hygrometers, catalystsCobalt(II) bromide hydrate is mainly used as a catalyst in organic synthesis, like C-metal, C-heteroatom and C-C bond formations, addition to unsaturated bonds, cyclizations, polymerization, oxidations reductions and carbonylations. It reacts with ammonia to form coordination compound bromopentaamminecobalt(III) bromide. It reacts with triphenylphosphine and form complexes of cobalt(II) bromide, which used as a catalyst in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 47°C to 48°C (decomposition)
Color Purple-Red
Physical Form Crystalline Granules
Quantity 50 g
Solubility Information Soluble in water.
IUPAC Name λ²-cobalt(2+) dibromide
Formula Weight 218.75 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Red-purple crystalline granules
Density 2.46 g/cm3
Chemical Name or Material Cobalt(II) bromide hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'