missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) bromide hydrate

Product Code. 10023913 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10023913 500 g Plastic bottle 500g
10297660 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10023913 Supplier Thermo Scientific Acros Supplier No. 318545000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85017-77-2 | Br2Co | 218.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85017-77-2
Molecular Formula Br2Co
Molecular Weight (g/mol) 218.74
MDL Number MFCD00016017
InChI Key BZRRQSJJPUGBAA-UHFFFAOYSA-L
Synonym cobalt ii bromide hydrate, acmc-20alea, br2co.h2o, cobaltous bromide hydrate, dibromocobalt-water 1/1, cobalt ii bromide hexahydrate, cobalt bromide cobr2 ,hydrate 9ci
PubChem CID 16212115
IUPAC Name λ²-cobalt(2+) dibromide
SMILES [Co++].[Br-].[Br-]

Specifications

Color Violet
Physical Form Crystals
Quantity 500 g
Linear Formula CoBr2·xH2O
Solubility Information Solubility in water: soluble in water
Formula Weight 218.75
Percent Purity 17.0 to 20.3% (Co)
Packaging Plastic bottle
Chemical Name or Material Cobalt(II) bromide hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.