missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) bromide, anhydrous, 97%

Product Code. 11352618
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352618 250 g 250g
11362618 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352618 Supplier Thermo Scientific Alfa Aesar Supplier No. 023149.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-43-7 | Br2Co | 218.741 g/mol

Cobalt(II) bromide is used as a catalyst in organic synthesis. It is used as a precursor in the production of ethylsulfanyl)porphyrazinato)cobalt(II), which gives the possibility of intermolecular ferromagnetic interactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-43-7
Molecular Formula Br2Co
Molecular Weight (g/mol) 218.741
MDL Number MFCD00016017
InChI Key BZRRQSJJPUGBAA-UHFFFAOYSA-L
Synonym cobalt bromide, cobalt ii bromide, cobalt bromide cobr2, cobalt dibromide, cobaltous bromide, hsdb 998, cobalt bromide ous
PubChem CID 24610
IUPAC Name dibromocobalt
SMILES [Co](Br)Br

Specifications

Melting Point 678°C (in N2)
Color Green
Physical Form Crystalline
Quantity 250 g
Merck Index 14,2435
Solubility Information Soluble in water,methanol,ethanol,acetone,methyl acetate Soluble in water,methyl acetate,ether,ethanol,methanol and acetone.
Formula Weight 218.75
Percent Purity 97%
Sensitivity Hygroscopic
Density 4.909 g/mL
Chemical Name or Material Cobalt(II) bromide, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.