missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) benzoate

Product Code. 11364979
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364979 250 g 250g
11344979 10 g 10g
11354979 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11364979 Supplier Thermo Scientific Alfa Aesar Supplier No. 087380.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 932-69-4 | C14H10CoO4 | 301.16 g/mol

Cobalt(II) benzoate is used as ingredients of many cosmetic products, industrial fields such as automobile, fuel, battery, paint, metallurgy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932-69-4
Molecular Formula C14H10CoO4
Molecular Weight (g/mol) 301.16
MDL Number MFCD00050769
InChI Key GAYAMOAYBXKUII-UHFFFAOYSA-L
Synonym cobalt 2+ dibenzoate, cobalt ii benzoate, cobalt dibenzoate, cobalt benzoate, benzoic acid, cobalt 2+ salt, benzoic acid, cobalt 2+ salt 2:1, cobaltous benzoate, dibenzoic acid cobalt ii salt
PubChem CID 164760
IUPAC Name cobalt(2+);dibenzoate
SMILES [Co++].[O-]C(=O)C1=CC=CC=C1.[O-]C(=O)C1=CC=CC=C1

Specifications

Melting Point 114°C to 116°C
Color Gray
Odor Odorless
Linear Formula Co(OOCC6H5)2
Quantity 250 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 301.16
Physical Form Crystalline
Chemical Name or Material Cobalt (II) benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.