missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) acetylacetonate, 99%

Product Code. 10286500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10286500 50 g Glass bottle 50g
10606164 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10286500 Supplier Thermo Scientific Acros Supplier No. 210640500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14024-48-7 | C10H14CoO4 | 257.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14024-48-7
Molecular Formula C10H14CoO4
Molecular Weight (g/mol) 257.15
MDL Number MFCD00000014
InChI Key UTYYEGLZLFAFDI-FDGPNNRMSA-L
IUPAC Name λ²-cobalt(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Co++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 165.0°C to 170.0°C
Color Purple
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5% (ex Co)
Packaging Glass bottle
Linear Formula (CH3COCH=C(CH3)O)2Co
Quantity 50 g
Beilstein 01, 783
Fieser 17,87
Solubility Information Solubility in water: 5g/L (20°C). Other solubilities: 7g/kg alcohol (20°C),173g/kg benzene (20°C)
Formula Weight 257.14
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Cobalt(II) acetylacetonate
Show More Show Less
compliance-icons compliance-icons
Identifiant produit
  • Cobalt(II) acetylacetonate
Mention d'avertissement
  • Danger
Catégorie de danger
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Acute toxicity Category 4
Mention de danger
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304-IF INHALED:.
  • P340-Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.