missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) acetate tetrahydrate, Co 24%

Product Code. p-4781861
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 11326648

Brand: Thermo Scientific Alfa Aesar 012227.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6147-53-1 | C4H14CoO8 | 249.081 g/mol

It is used as an industrial catalyst that allow paints and varnishes to harden. Also as a precursor to various oil drying agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6147-53-1
Molecular Formula C4H14CoO8
Molecular Weight (g/mol) 249.081
MDL Number MFCD00149569
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
Synonym cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
PubChem CID 61372
IUPAC Name cobalt(2+);diacetate;tetrahydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]

Specifications

Melting Point ∽140°C-4H2O
Color Red
Physical Form Crystals
Quantity 250 g
Assay Percent Range 24%
Beilstein 3731397
Solubility Information Soluble in water,alcohol,dilute acids
Formula Weight 249.08 (177.02 Anhydrous)
Odor Odorless
Refractive Index 1.542
Sensitivity Hygroscopic
Density 1.71 g/cm3
Chemical Name or Material Cobalt(II) acetate tetrahydrate, Co 24%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.