missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) acetate, anhydrous, Co 32% min., Thermo Scientific Chemicals

Catalog Number 11372608
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
71-48-7
C4H6CoO4
177.02
MFCD00008689
QAHREYKOYSIQPH-UHFFFAOYSA-L
cobalt ii acetate, cobalt diacetate, cobalt acetate, cobaltous diacetate, bis acetato cobalt, cobalt 2+ acetate, cobaltous acetate, acetic acid, cobalt 2+ salt, cobalt di acetate, cobalt 2+ diacetate
6277
CHEBI:85138
cobalt(2+);diacetate
[Co++].CC([O-])=O.CC([O-])=O
This item is not returnable. View return policy
71-48-7
C4H6CoO4
177.02
MFCD00008689
QAHREYKOYSIQPH-UHFFFAOYSA-L
cobalt ii acetate, cobalt diacetate, cobalt acetate, cobaltous diacetate, bis acetato cobalt, cobalt 2+ acetate, cobaltous acetate, acetic acid, cobalt 2+ salt, cobalt di acetate, cobalt 2+ diacetate
6277
CHEBI:85138
cobalt(2+);diacetate
[Co++].CC([O-])=O.CC([O-])=O

Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 298°C (decomposition)
Color Pink to Purple
Physical Form Powder
Quantity 25 g
Assay Percent Range 32% min. as Co
Solubility Information Soluble in waterSoluble in water,alcohol,dilute acids and pentyl acetate(tetrahydrate).
Formula Weight 177.02
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Cobalt(II) acetate, Anhydrous, Co 32% min.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'