missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) acetate, anhydrous, 98+%

Product Code. 11400827
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400827 250 g 250g
11490817 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400827 Supplier Thermo Scientific Alfa Aesar Supplier No. B23218.30
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 71-48-7 | C4H6CoO4 | 177.02 g/mol

Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.

Solubility
Soluble in water, alcohol and dilute acids.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-48-7
Molecular Formula C4H6CoO4
Molecular Weight (g/mol) 177.02
MDL Number MFCD00008689
InChI Key QAHREYKOYSIQPH-UHFFFAOYSA-L
Synonym cobalt ii acetate, cobalt diacetate, cobalt acetate, cobaltous diacetate, bis acetato cobalt, cobalt 2+ acetate, cobaltous acetate, acetic acid, cobalt 2+ salt, cobalt di acetate, cobalt 2+ diacetate
PubChem CID 6277
ChEBI CHEBI:85138
IUPAC Name cobalt(2+);diacetate
SMILES [Co++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 298°C (decomposition)
Odor Odorless
Linear Formula Co(OOCCH3)2
Quantity 250 g
UN Number UN3077
Sensitivity Hygroscopic
Merck Index 14,2433
Solubility Information Soluble in water,alcohol and dilute acids.
Formula Weight 177.02
Percent Purity ≥98%
Chemical Name or Material Cobalt(II) acetate, anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.