missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) 2-methoxyethoxide, 5% w/v in 2-methoxyethanol, 99% (m.b.), packaged under Ar in resealable ChemSeal bottles, Thermo Scientific Chemicals

Product Code. 11340008
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11340008 100 mL 100mL
11350008 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11340008 Supplier Thermo Scientific Alfa Aesar Supplier No. 042819.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used as a nano-materials, thin films and ferroelectric metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142600-61-1
Molecular Formula C6H14CoO4
Molecular Weight (g/mol) 209.107
MDL Number MFCD00798554
InChI Key BFBNUELICQBJBC-UHFFFAOYSA-N
Synonym cobalt ii 2-methoxyethoxide
PubChem CID 131873636
IUPAC Name cobalt(2+);2-methoxyethanolate
SMILES COCC[O-].COCC[O-].[Co+2]

Specifications

Packaging ChemSeal™ bottles
Linear Formula Co(OCH2CH2OCH3)2
Quantity 100 mL
UN Number UN3274
Sensitivity Air and moisture sensitive
Solubility Information Denatured by water. Miscible with alcohols,ketones and esters.
Formula Weight 209.11
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 5% w/v in 2-methoxyethanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Cobalt (II) 2-methoxyethoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.