missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) 2,4-pentanedionate

Product Code. 11367377
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367377 50 g 50g
11377377 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367377 Supplier Thermo Scientific Alfa Aesar Supplier No. 012537.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14024-48-7 | C10H14CoO4 | 257.15 g/mol

Cobalt(II) 2,4-pentanedionate is used as an important catalyst for organic reactions. It is also used as paint dryer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14024-48-7
Molecular Formula C10H14CoO4
Molecular Weight (g/mol) 257.15
MDL Number MFCD00000014
InChI Key UTYYEGLZLFAFDI-FDGPNNRMSA-L
Synonym Cobalt(II) acetylacetonate; Bis(acetylacetonato)cobalt(II)
IUPAC Name λ²-cobalt(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Co++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 165°C to 170°C
Color Blue-Purple
Odor Odorless
Quantity 50 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 257.14
Physical Form Crystalline
Chemical Name or Material Cobalt(II) 2,4-pentanedionate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) 2,4-pentanedionate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Cobalt(II) 2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.