missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Chloride Hexahydrate Cell Culture MP Biomedicals

Product Code. 11495102 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495102 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11495102 Supplier MP Biomedicals Supplier No. 0219464225

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7791-13-1
Quantity 25 g
Molecular Formula Cl2CoH12O6
MDL Number MFCD00149652
Synonym Cobalt Dichloride Hexahydrate, Cobaltous Chloride, Hexahydrate
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
Molecular Weight (g/mol) 237.92
Formula Weight 237.9
Grade Cell Culture
Density 1.920 g/mL
Chemical Name or Material Cobalt Chloride, Hexahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cobalt Chloride Hexahydrate, cell culture reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Cobalt Chloride Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.