missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Chloride Hexahydrate, >98%, (ACS Reagent Grade), MP Biomedicals

Product Code. 11420592 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11420592 25 g 25g
11430592 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 11420592

Brand: MP Biomedicals 0219403525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Quantity 25 g
Assay Percent Range >98%
Formula Weight 237.9
Grade ACS Reagent
Chemical Name or Material Cobalt Chloride Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.