missing translation for 'onlineSavingsMsg'
Learn More

Cobalt for AAS 1g/L HCl matrix, Fisher Chemical™

Product Code. 12973135 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Unit Size:
1000mL
100mL
500mL
This item is not returnable. View return policy

Product Code. 12973135

Brand: Fisher Chemical B 012 1000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Packaging Bottle
Physical Form Liquid
Concentration 1 g/L ±0.002 g/L
Type Flame Single-element Standard Solution
Solvent or Matrix 2% HCl
Quantity 1000 mL
Chemical Name or Material Cobalt
compliance-icons
Product Identifier
  • Single-element Standard Solution for AAS. Cobalt (Co) 1000mg/l in HCl 2%
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H350-May cause cancer.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P273-Avoid release to the environment.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.