missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coatomer Subunit Delta Antibody (OTI6A6), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70203PE
This item is not returnable.
View return policy
Description
Coatomer Subunit Delta Monoclonal specifically detects Coatomer Subunit Delta in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications
| Coatomer Subunit Delta | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| ARCN1 | |
| Full length recombinant protein of human Coatomer Subunit Delta (NP_001646) produced in HEK293T cells. | |
| 0.1 mL | |
| 372.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI6A6 | |
| Flow Cytometry | |
| Archain, archain 1, archain vesicle transport protein 1, coatomer delta subunit, coatomer protein complex, subunit delta, coatomer protein delta-COP, coatomer subunit delta, COPD, Delta-coat protein, delta-COP | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction