missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coatomer Subunit Delta Antibody (189), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43773
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
Coatomer Subunit Delta Monoclonal specifically detects Coatomer Subunit Delta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
| Coatomer Subunit Delta | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| ARCN1 | |
| Recombinant protein encompassing a sequence within the center region of human COPD. The exact sequence is proprietary. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 189 | |
| Western Blot 1:1000-1:20000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000 | |
| Archain, archain 1, archain vesicle transport protein 1, coatomer delta subunit, coatomer protein complex, subunit delta, coatomer protein delta-COP, coatomer subunit delta, COPD, Delta-coat protein, delta-COP | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 372 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction