Description
Heat exchanger that is coated in plastic to prevent damage to the heat exchanger caused by corrosive sample material.
Specifications
Specifications
| For Use With (Equipment) | KB PRO 720, KB PRO 1060 |
| Material | Aluminum |
| Type | Heat Exchanger |