missing translation for 'onlineSavingsMsg'
Learn More

Coal tar solution, contains 81.0-86.0% alcohol

Product Code. 11355897
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11355897 100 mL 100mL
11450600 500 mL 500mL
11345897 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11355897 Supplier Thermo Scientific Alfa Aesar Supplier No. 043671.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 8007-45-2

Coal tar is used for constructing roads, electricity generation and coking. It has been used as topical medical treatment for psoriasis and dandruff and is formulated as a soap or ointment. It is also used for other types of rashes (such as eczema, atopic dermatitis, chronic exudative dermatitis). Coal tar has mild antiseptic and anti-inflammatory effects, and relieves itching. It is also absorbed into the skin cells and is thought to inhibit DNA replication in these cells. This slows down cell division and stops the cells from multiplying excessively. In turn, this reduces the thickening and scaling of the skin and clears up psoriasis plaques. It also finds its application in surface coatings and sealants. Also used as a fuel in open hearth furnaces in the steel industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8007-45-2
MDL Number MFCD00146482

Specifications

Color Brown
Flash Point 13°C (55°F)
Odor Alcohol-like
Quantity 100 mL
UN Number UN1993
Concentration or Composition (by Analyte or Components) Contains 81.0 to 86.0% alcohol
Physical Form Liquid
Chemical Name or Material Coal tar solution

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.