Learn More
Coal tar solution, contains 81.0-86.0% alcohol
Description
Coal tar is used for constructing roads, electricity generation and coking. It has been used as topical medical treatment for psoriasis and dandruff and is formulated as a soap or ointment. It is also used for other types of rashes (such as eczema, atopic dermatitis, chronic exudative dermatitis). Coal tar has mild antiseptic and anti-inflammatory effects, and relieves itching. It is also absorbed into the skin cells and is thought to inhibit DNA replication in these cells. This slows down cell division and stops the cells from multiplying excessively. In turn, this reduces the thickening and scaling of the skin and clears up psoriasis plaques. It also finds its application in surface coatings and sealants. Also used as a fuel in open hearth furnaces in the steel industry
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown |
| Flash Point | 13°C (55°F) |
| Odor | Alcohol-like |
| Quantity | 2.5 L |
| UN Number | UN1993 |
| Solubility Information | Fully miscible in water. |
| Concentration or Composition (by Analyte or Components) | Contains 81.0 to 86.0% alcohol |
| Physical Form | Liquid |
| Chemical Name or Material | Coal tar solution |
For Manufacturing and Laboratory Use. Not for beverage use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.