missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Coagulation Tubes with Sodium Citrate Solution

Product Code. 13048702
Click to view available options
Capacity (Metric):
2 mL
3.5 mL
Closure Color:
Blue with Black Ring
Blue with White Ring
Unit Size:
Pack of 1200
This item is not returnable. View return policy

Product Code. 13048702

Brand: Greiner Bio-One 454332

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For routine hemostasis testing

 

  • 13 × 75mm tubes filled with buffered tri-sodium citrate solution
  • 3.2 % (0.109 mol/L) concentration
  • Mixing ratio is one part citrate to nine parts blood when tube is filled appropriately as indicated by fill mark on label
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • 18-month shelf life from date of manufacture
  • Blue nonscrew safety pull cap can be removed with a gentle pull

 

TRUSTED_SUSTAINABILITY

Specifications

Product Type Sodium Citrate Blood Collection Tube
Additive 3.2% Citrate Solution
Material PET
Capacity (Metric) 3.5 mL
Closure Color Blue with Black Ring
Tube Size 13 x 75 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.