missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor XIV/Protein C Rabbit anti-Human, Mouse, Rat, Clone: 3W3R7, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16427-100UL
This item is not returnable.
View return policy
Description
Coagulation Factor XIV/Protein C Monoclonal antibody specifically detects Coagulation Factor XIV/Protein C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| Coagulation Factor XIV/Protein C | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 3W3R7 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Anticoagulant protein C, APC, Autoprothrombin IIA, Blood coagulation factor XIV, EC 3.4.21, EC 3.4.21.69, PC, PROC1, protein C (inactivator of coagulation factors Va and VIIIa), vitamin K-dependent protein C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 300-461 of human Coagulation Coagulation Factor XIV/Protein C (P04070). | |
| 100 μg | |
| Apoptosis, Cancer | |
| 5624 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction