missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor XIV/Protein C Mouse anti-Human, Clone: PC 7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05113-0.025MG
This item is not returnable.
View return policy
Description
Coagulation Factor XIV/Protein C Monoclonal specifically detects Coagulation Factor XIV/Protein C in Human samples. It is validated for ELISA.
Specifications
| Coagulation Factor XIV/Protein C | |
| Monoclonal | |
| 1.0 mg/ml | |
| ELISA 1:500 | |
| Anticoagulant protein C, APC, Autoprothrombin IIA, Blood coagulation factor XIV, EC 3.4.21, EC 3.4.21.69, PC, PROC1, protein C (inactivator of coagulation factors Va and VIIIa), vitamin K-dependent protein C | |
| Recombinant form of human activated protein C (Drotrecogin alfa, Xigris) | |
| 0.025 mg | |
| Apoptosis, Cancer | |
| 5624 | |
| Store at 4°C. Do not freeze. | |
| IgG1 κ |
| ELISA | |
| PC 7 | |
| Unconjugated | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction