missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coagulation Factor XIV/Protein C Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Coagulation Factor XIV/Protein C |
---|---|
Clone | 390226 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Coagulation Factor XIV/Protein C Monoclonal specifically detects Coagulation Factor XIV/Protein C in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Coagulation Factor XIV/Protein C | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
P04070 | |
5624 | |
Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor XIV/Protein C Ala43-Pro461 Accession # P04070 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Coagulation Factor XIV/Protein C in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Factor VII, rhFactor X, rhFactor XI, rhThrombin, rhUPA, rhHGFA, rhKallikrein 1, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 13, 14, or 15 is observed. |
390226 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Anticoagulant protein C, APC, Autoprothrombin IIA, Blood coagulation factor XIV, EC 3.4.21, EC 3.4.21.69, PC, PROC, PROC1, Protein C, protein C (inactivator of coagulation factors Va and VIIIa), vitamin K-dependent protein C | |
PROC | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only