missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Coagulation Factor XI Antibody (Novo Nordisk patent anti-Factor VIII) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | Coagulation Factor XI |
|---|---|
| Clone | Novo Nordisk patent anti-Factor VIII |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30045668
|
Novus Biologicals™
NBP3-28739-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045503
|
Novus Biologicals™
NBP3-28739-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047919
|
Novus Biologicals™
NBP3-28739-100ug |
100 μg |
£257.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Coagulation Factor XI Monoclonal antibody specifically detects Coagulation Factor XI in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| Coagulation Factor XI | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Prostate Cancer | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| coagulation factor XI, EC 3.4.21, EC 3.4.21.27, FXIPlasma thromboplastin antecedent, MGC141891, PTA | |
| F8/ Factor VIII | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| Novo Nordisk patent anti-Factor VIII | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P00451 | |
| 2160 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title