missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Coagulation Factor XI Antibody (garadacimab) - IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28438-1mg
This item is not returnable.
View return policy
Description
Coagulation Factor XI Monoclonal antibody specifically detects Coagulation Factor XI in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Coagulation Factor XI | |
| Monoclonal | |
| Unconjugated | |
| P00748 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| garadacimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| coagulation factor XI, EC 3.4.21, EC 3.4.21.27, FXIPlasma thromboplastin antecedent, MGC141891, PTA | |
| F12/ Factor XII | |
| 1 mg | |
| Prostate Cancer | |
| 2160 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction