missing translation for 'onlineSavingsMsg'
Learn More

Coagulation Factor X Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18406979 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406979 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406979 Supplier Novus Biologicals Supplier No. NBP299698MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Coagulation Factor X Polyclonal antibody specifically detects Coagulation Factor X in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Host Species Rabbit
Quantity 0.1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.