missing translation for 'onlineSavingsMsg'
Learn More

Coagulation Factor X Antibody (3F2), Novus Biologicals™

Rabbit Monoclonal Antibody

£260.00 - £406.00

Specifications

Antigen Coagulation Factor X
Clone 3F2
Dilution ELISA, Immunohistochemistry 1:50-1:200
Applications ELISA, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043931
View Documents
Novus Biologicals
NBP3-26595-50ul
50 μL
£260.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043488
View Documents
Novus Biologicals
NBP3-26595-100ul
100 μL
£406.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Coagulation Factor X Monoclonal antibody specifically detects Coagulation Factor X in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications

Specifications

Coagulation Factor X
ELISA, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
PBS, pH 7.4, 150mM NaCl and 50% glycerol
2159
IgG
Affinity purified
3F2
ELISA, Immunohistochemistry
Unconjugated
Rabbit
Human
coagulation factor X, EC 3.4.21, EC 3.4.21.6, factor Xa, FX, FXA, prothrombinase, Stuart factor, Stuart-Prower factor
A synthesized peptide derived from Human Coagulation Factor X [UniProt P00742]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.