missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor X Antibody (058), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89784AF488
This item is not returnable.
View return policy
Description
Coagulation Factor X Monoclonal specifically detects Coagulation Factor X in Human samples. It is validated for Sandwich ELISA Detection.
Specifications
| Coagulation Factor X | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 2159 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 58 | |
| Sandwich ELISA Detection | |
| coagulation factor X, EC 3.4.21, EC 3.4.21.6, factor Xa, FX, FXA, prothrombinase, Stuart factor, Stuart-Prower factor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Coagulation Factor X (NP_000495.1; Met1-Lys488). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction