missing translation for 'onlineSavingsMsg'
Learn More

Coagulation Factor VII Antibody (RFF-VII/1) [Janelia Fluor™ 585], Novus Biologicals™

Product Code. 30177886
Change view
Click to view available options
Quantity:
0.25 mL
Unit Size:
0.25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30177886 0.25 mL 0.25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30177886 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP145150JF585

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Coagulation Factor VII Monoclonal antibody specifically detects Coagulation Factor VII in Human samples. It is validated for ELISA,Western Blot,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coagulation Factor VII
Applications ELISA, Western Blot, Functional Assay
Classification Monoclonal
Clone RFF-VII/1
Conjugate Janelia Fluor 585
Formulation 50mM Sodium Borate
Gene Alias coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA
Host Species Mouse
Immunogen Purified human Coagulation Factor VII
Purification Method Protein A purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 2155
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.