missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor VII Antibody (RFF-VII/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-45150
This item is not returnable.
View return policy
Description
Coagulation Factor VII Monoclonal specifically detects Coagulation Factor VII in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Coagulation Factor VII | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Functional | |
| coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| VII. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Functional Assay | |
| RFF-VII/1 | |
| Unconjugated | |
| P08709 | |
| F7 | |
| Purified human Coagulation Factor VII | |
| 0.25 mg | |
| Cardiovascular Biology | |
| 2155 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction