missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor VII Antibody (RFF-VII/1), Alexa Fluor™ 405, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Coagulation Factor VII Monoclonal antibody specifically detects Coagulation Factor VII in Human samples. It is validated for Western Blot, ELISA, Functional Assay
Specifications
Specifications
| Antigen | Coagulation Factor VII |
| Applications | Western Blot, ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | RFF-VII/1 |
| Conjugate | Alexa Fluor 405 |
| Formulation | 50mM Sodium Borate |
| Gene Accession No. | P08709 |
| Gene Alias | coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA |
| Host Species | Mouse |
| Immunogen | Purified human Coagulation Factor VII |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?