missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor VII Antibody (F7/3511) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08251
This item is not returnable.
View return policy
Description
Coagulation Factor VII Monoclonal specifically detects Coagulation Factor VII in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Coagulation Factor VII | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry-Paraffin : 1-2 ug/mL, Protein Array | |
| coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA | |
| Recombinant fragment (around aa 366-466) of human Coagulation Factor VII protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin), Peptide Array | |
| F7/3511 | |
| Unconjugated | |
| 10mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2155 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction