missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Coagulation Factor III/Tissue Factor Antibody (tisotumab), Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | Coagulation Factor III/Tissue Factor |
|---|---|
| Clone | tisotumab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30048508
|
Novus Biologicals™
NBP3-28700-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30048682
|
Novus Biologicals™
NBP3-28700-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045748
|
Novus Biologicals™
NBP3-28700-100ug |
100 μg |
£247.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Coagulation Factor III/Tissue Factor Monoclonal antibody specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| Coagulation Factor III/Tissue Factor | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Cancer | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
| F3/ Factor III/ Tissue Factor/ CD142 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| tisotumab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P13726 | |
| 2152 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title