missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor III/Tissue Factor Antibody (SN20-16), Novus Biologicals™
Rabbit Monoclonal Antibody
£149.00 - £403.00
Specifications
| Antigen | CoagulationFactorIII/TissueFactor |
|---|---|
| Clone | SN20-16 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:500 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18118722
|
Novus Biologicals
NBP2-67731-25UL |
25 ul |
£149.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18041736
|
Novus Biologicals
NBP2-67731 |
100 μL |
£403.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Coagulation Factor III/Tissue Factor Monoclonal specifically detects Coagulation Factor III/Tissue Factor in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CoagulationFactorIII/TissueFactor | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Cancer | |
| CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
| F3 | |
| IgG | |
| Protein A purified |
| SN20-16 | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:500 | |
| Unconjugated | |
| Rabbit | |
| TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
| 2152 | |
| Synthetic peptide within human Coagulation Factor III/Tissue Factor aa 30-70. (SwissProt: P13726 Human; SwissProt: P20352 Mouse; SwissProt: P42533 Rat) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title