Learn More
Coagulation Factor III/Tissue Factor Antibody (CL3805) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61640F
Product Code. 30295574
  • £558.00 / 0.10mL
Description
Coagulation Factor III/Tissue Factor Monoclonal antibody specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Coagulation Factor III/Tissue Factor
Monoclonal
FITC
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor
This antibody was developed using a recombinant protein derived from P13726, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
2152
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL3805
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified