missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor III/Tissue Factor Antibody (001), mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Coagulation Factor III/Tissue Factor Monoclonal antibody specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for ELISA
Specifications
Specifications
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Coagulation Factor III/Tissue Factor (Uniprot#: P13726; Met1-Glu251). |
| Purification Method | Protein A purified |
| Regulatory Status | RUO |
| Research Discipline | Cancer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?