missing translation for 'onlineSavingsMsg'
Learn More

Coagulation Factor II/Thrombin Mouse anti-Human, Clone: 22, Novus Biologicals™

Product Code. 18111709 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.10mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18111709 0.025 mg 25µg
18612957 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18111709 Supplier Novus Biologicals Supplier No. NBP2413810.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Coagulation Factor II/Thrombin Monoclonal specifically detects Coagulation Factor II/Thrombin in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coagulation Factor II/Thrombin
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
Clone 22
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Sandwich ELISA 1:100-1:2000, Sandwich ELISA Capture 1:100-1:2000
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl
Gene Alias Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin receptor-like 2
Host Species Mouse
Immunogen Thrombin-antithrombin (TAT) complexes
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2147
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.