missing translation for 'onlineSavingsMsg'
Learn More

Coagulation Factor II/Thrombin Antibody (046), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18352741 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352741 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18352741 Supplier Novus Biologicals Supplier No. NBP290542AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Coagulation Factor II/Thrombin Monoclonal specifically detects Coagulation Factor II/Thrombin in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coagulation Factor II/Thrombin
Applications ELISA
Classification Monoclonal
Clone 46
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin receptor-like 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Coagulation Factor II/Thrombin (NP_034298.1; Met1-Gly618).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2147
Test Specificity No cross-reactivity in ELISA with: Mouse KLK1 Mouse KLK11
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.