missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor II/Thrombin Antibody (046), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90542AF350
This item is not returnable.
View return policy
Description
Coagulation Factor II/Thrombin Monoclonal specifically detects Coagulation Factor II/Thrombin in Mouse samples. It is validated for ELISA.
Specifications
| Coagulation Factor II/Thrombin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin receptor-like 2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Coagulation Factor II/Thrombin (NP_034298.1; Met1-Gly618). | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, GPCR, Signal Transduction | |
| 2147 | |
| Mouse | |
| Purified |
| ELISA | |
| 46 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Mouse KLK1 Mouse KLK11 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction