missing translation for 'onlineSavingsMsg'
Learn More

Coactosin-like Protein 1/CotL1 Antibody [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30730959 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30730959 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30730959 Supplier Novus Biologicals Supplier No. AF7865MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Coactosin-like Protein 1/CotL1 Polyclonal antibody specifically detects Coactosin-like Protein 1/CotL1 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coactosin-like Protein 1/CotL1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias CLPFLJ43657, coactosin-like 1 (Dictyostelium), coactosin-like protein, MGC19733
Host Species Sheep
Immunogen E. coli-derived recombinant human Coactosin-like Protein 1/COTL1, Ala2-Glu142, Accession # Q14019
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23406
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.