missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COA7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100413
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total C1orf163.
Required for assembly of mitochondrial respiratory chain complex I and complex IV.
Specifications
| COA7 | |
| Polyclonal | |
| Unconjugated | |
| COA7 | |
| 2010305A19Rik; AI451324; beta-lactamase hcp-like protein; C1orf163; COA7; Cytochrome c oxidase assembly factor 7; cytochrome c oxidase assembly factor 7 (putative); D4Ertd796e; hcp beta-lactamase-like protein C1orf163; hcp beta-lactamase-like protein C1orf163 homolog; RESA1; respiratory chain assembly 1; respiratory chain assembly factor 1; RGD1305274; Sel1 repeat containing 1; sel1 repeat-containing protein 1; Selrc1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 65260, 69893 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q921H9, Q96BR5 | |
| COA7 | |
| A synthesized peptide derived from human COA7(Accession Q96BR5), corresponding to amino acid residues P103-L153. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction