missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572018
This item is not returnable.
View return policy
Description
COA4 Polyclonal Antibody for Western Blot
Putative COX assembly factor.
Specifications
| COA4 | |
| Polyclonal | |
| Unconjugated | |
| COA4 | |
| 5330414O08Rik; AI115477; AW047259; CHCHD8; CMC3; Coa4; coiled-coil-helix-coiled-coil-helix domain containing 8; coiled-coil-helix-coiled-coil-helix domain-containing protein 8; cytochrome c oxidase assembly factor 4; cytochrome c oxidase assembly factor 4 homolog; cytochrome c oxidase assembly factor 4 homolog, mitochondrial; E2ig2; E2-induced gene 2 protein; RGD1562228 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 51287 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.45 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9NYJ1 | |
| COA4 | |
| KLH conjugated synthetic peptide between 56-84 amino acids from the C-terminal region of human CHCH8. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction