Learn More
Invitrogen™ CoA Synthase Recombinant Rabbit Monoclonal Antibody (7K2O6)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542635
Description
Positive test controls include: HT-29, 293T. The target is usually found in the following locations: Cytoplasm, Mitochondrion matrix. Immunogen sequence: RVCVIDAAVL LEAGWQNLVH EVWTAVIPET EAVRRIVERD GLSEAAAQSR LQSQMSGQQL VEQSHVVLST LWEPHITQRQ VEKAWALLQK RIPKTHQALD.
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. COASY is a bifunctional enzyme that catalyzes the 2 last steps in CoA synthesis. These activities are performed by 2 separate enzymes, phosphopantetheine adenylyltransferase and dephospho-CoA kinase, in prokaryotes.
Specifications
| CoA Synthase | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13057 | |
| COASY | |
| A synthetic peptide corresponding to a sequence within amino acids 465-564 of human CoA synthase (COASY) (Q13057). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 7K2O6 | |
| Unconjugated | |
| COASY | |
| 1300003G02Rik; bifunctional coenzyme A synthase; bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase; CoA synthase; Coasy; Coenzyme A synthase; Dephospho-CoA kinase; Dephospho-CoA pyrophosphorylase; Dephosphocoenzyme A kinase; DPCK; DPCOAK; NBIA6; NBP; nucleotide binding protein; Pantetheine-phosphate adenylyltransferase; Phosphopantetheine adenylyltransferase; phosphopantetheine adenylyltransferase / dephosphocoenzyme A kinase; POV-2; PPAT; PSEC0106; Ukr1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 80347 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.